missing translation for 'onlineSavingsMsg'
Learn More

Podoplanin Antibody (56F7), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-59445AF594

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18216150

  • 562.62€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

Podoplanin Monoclonal antibody specifically detects Podoplanin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Especificaciones

Especificaciones

Podoplanin
Monoclonal
Alexa Fluor 594
36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha
Recombinant human Podoplanin (99-207aa) purified from E. coli
0.1 mL
Core ESC Like Genes, Signal Transduction, Stem Cell Markers
10630
Store at 4°C in the dark.
IgG1 κ
Western Blot, Flow Cytometry, ELISA, Immunofluorescence
56F7
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales