missing translation for 'onlineSavingsMsg'
Learn More

PMS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 18164-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16818224

  • 109.00€ / 20 microlitros

Envío estimado: 22-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This antibody is specific to PMS2. The calculated molecular weight of PMS2 is 96 kDa, but the modified PMS2 is about 117 kDa.

PMS1 protein homologue 2 (PMS2) is a DNA mismatch repair protein. Defects in PMS2 are a cause of hereditary non-polyposis cancer (HNPCC). Human PMS2 is an amino acid protein of 862 amino acids.
Especificaciones

Especificaciones

PMS2
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P54278, P54279
HNPCC4, PMS1 protein homolog 2, PMS2, PMS2CL, PMSL2
Rabbit
IgG
20 μL
-20°C
Primary
18861, 288479, 5395
Western Blot
Unconjugated
PMS2
Liquid
Pms2
PMS2 Fusion Protein Ag12651
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Human, Mouse
Videos
SDS
Documentos
Certificados