missing translation for 'onlineSavingsMsg'
Learn More

PMPCA Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA548135

Detalles adicionales : Peso : 0.01000kg

Código de producto. 15642435

  • 339.00€ / 400 microlitros


Fecha de envío estimada
del proveedor 23-06-2020
Añadir a la cesta

Descripción

Descripción

PMPCA cleaves presequences (transit peptides) from mitochondrial protein precursors.
Especificaciones

Especificaciones

PMPCA
Polyclonal
PBS with 0.09% sodium azide
Q10713
Mitochondrial-processing peptidase subunit alpha, Alpha-MPP, P-55, PMPCA, INPP5E, KIAA0123, MPPA
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23203
Flow Cytometry, Western Blot
Unconjugated
PMPCA
Liquid
PMPCA
KLH conjugated synthetic peptide between 454-483 amino acids from the C-terminal region of human PMPCA.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Videos
Documentos