Learn More
ERLAB PMMA Cable Ducts
Click to view available options
Para utilizar con (equipo):
Campanas extractoras y carcasas
Conducto de aire de escape y auxiliar
Tamaño de la unidad:
cada uno
Descripción
- Cable ducts allow clean, safe installation of electrical cables in the hood workspace
- Provide chemical resistance and visual inspection of cables
- Avoid cable clutter and protect against mechanical wear and contamination
- Mounted on the right side of the hood for ergonomic access and optimal cable management
- Improve airflow fluidity by reducing obstruction caused by cables entering directly through the chassis
- Ideal for configurations involving multiple instruments, sensors or monitoring devices
Especificaciones
Especificaciones
| Para utilizar con (equipo) | Conducto de aire de escape y auxiliar |
| Tipo de producto | Upper Duct Transition Adapter |