Learn More
PLXNA3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA566227
Detalles adicionales : Peso : 0.01000kg
Descripción
Immunogen sequence: SSLDAGSRVTV TVRDSECQFV RRDAKAIVCI SPLSTLGPSQ APITLAIDRA NISSPGLIYT YTQDPTVTRL EPTWSIINGS TA Highest antigen sequence identity to the following orthologs - mouse 84%, rat 86%.
PLXNA3 Polyclonal antibody specifically detects PLXNA3 in Human samples. It is validated for Immunocytochemistry, Western Blot.Especificaciones
PLXNA3 | |
Polyclonal | |
Unconjugated | |
Plxna3 | |
6.3; HSSEXGENE; mKIAA4078; PlexA3; plexin 3; Plexin 4, SEX, homolog to the cMet/HGF receptors; plexin A3; plexin-4; Plexin-A3; Plxa3; Plxn3; Plxn4; Plxna3; Semaphorin receptor SEX; SEX; Sex chromosome X transmembrane protein of HGF receptor family 3; SEXPLEXIN-A3; XAP-6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55558 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P51805 | |
Plxna3 | |
Recombinant Human PLXNA3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only