Learn More
Fisherbrand™ Plug Without Cable Electrode
Product Code. 11756199 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Length (Metric) Cable:
5 m
Unit Size:
Each
Product Code. Length (Metric) Cable Unit Size
11756199 5 m Each
1 options
Product Code. 11756199
  • ONLINE EXCLUSIVE PRICE
    27.45€
    17.77€ / paquete de 1
    Ahorro 9.68€
    35% Descuento
Fecha estimada de envoi: 07-01-2027
Add to basket
Add to basket
This durable coaxial cable for S7 or S8 head electrodes is ideal for connecting screw terminals of pH regulators.
  • Plug for connection to screw terminals of pH regulators
  • For combined electrodes
  • Plug without cable electrode
  • S7 cable unplug 5M
Specifications
Connector Types S7
Description Plug Without Cable Electrode
Type Plug
Length (Metric) Cable 5 m