missing translation for 'onlineSavingsMsg'
Learn More

PLEKHH3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 28154-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16816025

  • 319.00€ / 150 microlitros

Envío estimado: 27-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

28154-1-AP detects the isoforms of PLEKHH3 protein around 80-85 kDa in SDS-PAGE.

PLEKHH3, a gene related to endothelial tight junction, is associated with obstructive sleep apnea (OSA) and angiogenesis. It has been reported that the expression of PLEKHH3 protein was increased in the OSA patients. PLEKHH3 may play a key role in OSA and angiogenesis by specifically regulating the endothelial cells formation of tight junctions.
Especificaciones

Especificaciones

PLEKHH3
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q7Z736, Q8VCE9
FLJ21019, PLEKHH3
Rabbit
IgG
150 μL
-20°C
Primary
217198, 79990
Western Blot
Unconjugated
PLEKHH3
Liquid
PLEKHH3
PLEKHH3 Fusion Protein Ag28001
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
Documentos
Certificados