missing translation for 'onlineSavingsMsg'
Learn More

PLEKHH2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA567446

Detalles adicionales : Peso : 0.01000kg

Código de producto. 15900105

  • 410.00€ / 100 microlitros


Fecha de envío estimada
del proveedor 03-07-2020
Añadir a la cesta

Descripción

Descripción

PLEKHH2 contains 1 FERM domain, 1 MyTH4 domain and 2 PH domains. It is a Single-pass membrane protein. The exact function of PLEKHH2 remains unknown.
Especificaciones

Especificaciones

PLEKHH2
Polyclonal
Unconjugated
PLEKHH2
Liquid
PLEKHH2
Recombit Human PLEKHH2
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8IVE3
KIAA2028
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
130271
Videos
Documentos

For Research Use Only