Learn More
PLEKHH1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA562523
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: KSHNQRLVEQ VGSLQDALEA IQIAPSRKLL VPPYGAAEQD SVPSEPGIQP MGQDSGSQAQ GLKAAVLAPS PGALQSKDSV SE Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 54%.
PLEKHH1 Polyclonal antibody specifically detects PLEKHH1 in Human samples. It is validated for Immunohistochemistry.Especificaciones
PLEKHH1 | |
Polyclonal | |
Unconjugated | |
PLEKHH1 | |
KIAA1200; PH domain-containing family H member 1; pleckstrin homology domain containing, family H (with MyTH4 domain) member 1; pleckstrin homology domain-containing family H member 1; pleckstrin homology, MyTH4 and FERM domain containing H1; PLEKHH1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57475 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9ULM0 | |
PLEKHH1 | |
Recombinant protein corresponding to Human PLEKHH1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only