Learn More
Plectin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA555280
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: LTGLSLLPLS EKAARARQEE LYSELQARET FEKTPVEVPV GGFKGRTVTV WELISSEYFT AEQRQELLRQ FRTGKVTVEK VIKILIT Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 91%.
Plectin Polyclonal antibody specifically detects Plectin in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Especificaciones
Plectin | |
Polyclonal | |
Unconjugated | |
Plec | |
AA591047; AU042537; EBS1; EBSMD; EBSND; EBSO; EBSOG; EBSPA; HD1; Hemidesmosomal protein 1; LGMD2Q; PCN; Plec; PLEC1; PLEC1b; Plectin; plectin 1; plectin 1, intermediate filament binding protein 500kDa; plectin-1; Plectin-6; Pltn | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5339 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15149 | |
Plec | |
Recombinant protein corresponding to Human Plectin | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only