missing translation for 'onlineSavingsMsg'
Learn More

PLCE1 Rabbit anti-Human, Polyclonal, Thermo Scientific™ Pierce™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA553334

Detalles adicionales : Peso : 0.02000kg

Código de producto. 15791271

  • 376.00€ / 100 microlitros


Fecha de envío estimada
del proveedor 30-06-2020
Añadir a la cesta

Descripción

Descripción

Immunogen sequence: NSQEETLEFV ADYSGQDNFL QRVGQNGLKN SEKESTVNSI FQVIRSCNRS LETDEEDSPS EGNSSRKSSL KDKSRWQFII GDLLDSDNDI FEQSKEYDSH GSEDSQKAFD HGTELIPWYV LSIQADVHQF LLQGATVIHY DQD Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.

Especificaciones

Especificaciones

PLCE1
Unconjugated
Q9P212
PLCE1
Recombinant protein corresponding to Human PLCE1
100μL
Store at 4°C short term. Store at −20°C long term. Avoid freeze/thaw cycles.
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
100μL purified antibody (Lot-Specific) in PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Liquid
Rabbit
IgG
RUO
Primary
51196
Videos
Documentos

Research Use Only