missing translation for 'onlineSavingsMsg'
Learn More

PLCB3 Mouse anti-Human, Mouse, Rat, Clone: 1B8B3, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66668-1-IG-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16818765

  • 319.00€ / 150 microlitros

Envío estimado: 29-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

PLCB3 encodes a member of the phosphoinositide phospholipase C beta enzyme family that catalyze the production of the secondary messengers diacylglycerol and inositol 1,4,5-triphosphate from phosphatidylinositol in G-protein-linked receptor-mediated signal transduction. Alternative splicing results in multiple transcript variants.
Especificaciones

Especificaciones

PLCB3
1B8B3
Unconjugated
PLCB3
Liquid
Plcb3
PLCB3 Fusion Protein Ag15845
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P51432, Q01970, Q99JE6
FLJ37084, Phospholipase C beta 3, PLC beta 3, PLCB3
Mouse
IgG1
150 μL
-20°C
Primary
18797, 29322, 5331
Videos
SDS
Documentos
Certificados