Learn More
DWK Life Sciences™ Plate Holder, for glass ceramic laboratory protection plate
Product Code. 11772909 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric):
135 mm
155 mm
175 mm
Unit Size:
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
11772909 175 mm Pack of 5
11762909 155 mm Pack of 5
11752909 135 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11772909 Supplier DWK Life Sciences™ Supplier No. 290785809
  • ONLINE EXCLUSIVE PRICE
    275.00€
    239.64€ / paquete de 5
    Ahorro 35.36€
    12% Descuento
Fecha estimada de envoi: 25-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Ideal holder for glass ceramic plates for mounting on tripods.
In the laboratory, solutions are often heated. This step can be carried out using different techniques, e.g. in a water or sand bath, with a heating hood or classically using a Bunsen burner. If the Bunsen burner is used as the heat source, it is recommended to use a tripod or square foot and a corresponding glass ceramic plate. The vessel to be heated is placed on the glass ceramic plate with the Bunsen burner underneath. Contamination of the glass ceramic plate can be removed with the cleaning scraper.
  • Made from heat-resistant chrome-nickel steel with
Specifications
Material Chrome-nickel Steel
Diameter (Metric) 175 mm
For Use With (Application) Plate holder for glass ceramic plates; Heating; Suitable for mounting on tripods
No. per Pack 5/Pk.