Learn More
Brady™ Plastic Coated Steel Cable
Product Code. 11786685 Shop All Brady Products
Change view
Click to view available options
Length (Metric):
1.83 m
3.05 m
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11786685 3.05 m Each
11766685 1.83 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11786685 Supplier Brady™ Supplier No. 800115
  • ONLINE EXCLUSIVE PRICE
    38.75€
    33.79€ / paquete de 1
    Ahorro 4.96€
    12% Descuento
Fecha estimada de envoi: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Additional Plastic Coated Steel Cable for Cable Lockout Device.
  • Additional Cable for Cable Lockout Device.
Specifications
Product Type Cable
Alternate Part Number CABLE-3 M
Color Red
Description Additional Cable 3 m
Diameter (English) 0.19 in.
Diameter (Metric) 4.76 mm
Length (English) 10.01 ft.
Length (Metric) 3.05 m
Quantity 1 Ea.