Learn More
Epredia™ Lab Vision™ Plasma Cell Ab-1 Mouse Monoclonal Antibody, 50μg/mL, BSA and azide
Provide accurate, reproducible results with the Epredia™ Plasma Cell Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia™ MS-488-P0
Detalles adicionales : Peso : 1.00000kg
Descripción
Host Species: Mouse
Clone: LIV3G11
Isotype: IgG2a
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: Not determined
Positive Control: Tonsil or lymph node
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin only)
Note:
USA: RUO; Int'l: RUO
Especificaciones
Plasma Cell Ab-1 | |
Monoclonal | |
50μg/mL | |
Mouse | |
RUO | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
LIV3G11 | |
Unconjugated | |
100 μL | |
Hematopoietic | |
Human | |
IgG2a |