missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKNOX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20352-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
PREP-2 (Pbx-regulating protein-2), also known as PBX/knotted 1 homeobox 2 or PKNOX2, is a widely expressed protein belonging to the TALE (three amino acid loop extension)/MEIS family. PREP-2 is a DNA-binding protein that forms stable complexes with Pbx proteins. It is highly homologous to the related protein PREP-1, but displays a more restricted tissue distribution and a higher DNA-dissociation rate. Like PREP-1, PREP-2 forms a heterodimer with Pbx 1. The PREP-2-Pbx 1 dimer is relocated to the nucleus where it associates with HoxB1 to form a ternary complex. In contrast with PREP-1, which acts to increase transcriptional activation in this ternary complex, PREP-2 leads to a slight decrease in transcriptional activity of the ternary complex. Multiple isoforms exist for PREP-2, localizing to the nucleus or cytoplasm. Cytoplasmic isoforms are believed to colocalize with F-actin, G-actin and tubulin/microtubules.Especificaciones
PKNOX2 | |
Polyclonal | |
Unconjugated | |
PKNOX2 | |
Homeobox protein PKNOX2, Homeobox protein PREP 2, PBX/knotted 1 homeobox 2, PBX/knotted homeobox 2, PKNOX2, PREP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
208076, 63876, 680549 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.6 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BG99, Q96KN3 | |
PKNOX2 | |
PKNOX2 Fusion Protein Ag14195 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |