missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKIG Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA568010
Detalles adicionales : Peso : 0.01000kg
Descripción
Extremely potent competitive inhibitor of cAMP-dependent protein kinase activity, this protein interacts with the catalytic subunit of the enzyme after the cAMP-induced dissociation of its regulatory chains.Especificaciones
PKIG | |
Polyclonal | |
Unconjugated | |
PKIG | |
PKI-gamma; cAMP-dependent protein kinase inhibitor gamma; protein kinase (cAMP-dependent, catalytic) inhibitor gamma | |
Rabbit | |
Affinity chromatography | |
RUO | |
11142 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9Y2B9 | |
PKIG | |
Synthesized peptide derived from human PKIG. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |