missing translation for 'onlineSavingsMsg'
Learn More

PKC alpha Antibody (MC5), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Bio-Techne NB600-201AF350

Additional Details : Peso : 0.00970kg

Product Code. 18214374

  • 549.66€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Description

Description

PKC alpha Monoclonal specifically detects PKC alpha in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Fish, Rabbit, Sheep, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

PKC alpha
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
PRKCA
Purified bovine brain Protein Kinase C
Primary
This antibody is capable of detecting PKC beta as well.
Store at 4°C in the dark.
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
MC5
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
AAG6, aging-associated gene 6, EC 2.7.11, EC 2.7.11.13, MGC129901, PKC-A, PKC-alpha, PKCAMGC129900, PRKACA, protein kinase C alpha type, protein kinase C, alpha
Mouse
0.1 mL
5578
Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Fish, Rabbit, Sheep
IgG2a Kappa
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers