Learn More
PITPNM2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA551929
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: PKDITKWSSN DLMDKIESPE PEDTQDGLYR QGAPEFRVAS SVEQLNIIED EVSQPLAAPP SKIHVLLLVL HGGTILDTGA GDPSSKKGDA NTIANVFDTV MRVHYPSALG RLAIRLVPCP PVCSDAFALV SN Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
PITPNM2 Polyclonal antibody specifically detects PITPNM2 in Human samples. It is validated for Immunohistochemistry.Especificaciones
PITPNM2 | |
Polyclonal | |
Unconjugated | |
PITPNM2 | |
KIAA1457; Membrane-associated phosphatidylinositol transfer protein 2; NIR3; NIR-3; phosphatidylinositol transfer protein membrane associated 2; phosphatidylinositol transfer protein, membrane associated 2; phosphatidylinositol transfer protein, membrane-associated 2; PITPnm 2; PITPNM2; Pyk2 N-terminal domain-interacting receptor 3; RDGB2; RDGBA2; retinal degeneration B alpha 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57605 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BZ72 | |
PITPNM2 | |
Recombinant protein corresponding to Human PITPNM2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only