missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PILRA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA566000
Detalles adicionales : Peso : 0.01000kg
Descripción
Immunogen sequence: ALSSSTSPRAP PSHRPLKSPQ NETLYSVLK Highest antigen sequence identity to the following orthologs - mouse 50%, rat 57%.
PILRA Polyclonal antibody specifically detects PILRA in Human samples. It is validated for Immunohistochemistry.Especificaciones
PILRA | |
Polyclonal | |
Unconjugated | |
PILRA | |
AV021745; cell surface receptor FDF03; FDF03; Inhibitory receptor PILR-alpha; paired immunoglobin like type 2 receptor alpha; paired immunoglobin-like type 2 receptor alpha; paired immunoglobulin-like type 2 receptor alpha; Pilra | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29992 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UKJ1 | |
PILRA | |
Recombinant Human PILRA | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only