missing translation for 'onlineSavingsMsg'
Learn More

PIGP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA548800

Detalles adicionales : Peso : 0.01000kg

Código de producto. 15663095

  • 339.00€ / 400 microlitros


Fecha estimada de envío
31-08-2020
Añadir a la cesta

Descripción

Descripción

Part of the complex catalyzing the transfer of N- acetylglucosamine from UDP-N-acetylglucosamine to phosphatidylinositol, the first step of GPI biosynthesis.
Especificaciones

Especificaciones

PIGP
Polyclonal
PBS with 0.09% sodium azide
P57054
Phosphatidylinositol N-acetylglucosaminyltransferase subunit P, Down syndrome critical region protein 5, Down syndrome critical region protein C, Phosphatidylinositol-glycan biosynthesis class P protein, PIG-P, PIGP, DCRC, DSCR5, DSCRC
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51227
Western Blot
Unconjugated
PIGP
Liquid
PIGP
KLH conjugated synthetic peptide between 2-30 amino acids from the N-terminal region of human PIGP.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Videos
Documentos