Learn More
Thermo Scientific™ Pierce™ Iodination Tubes
Product Code. 11864614 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 Tubes
Unit Size:
Pack of 10
Product Code. Quantity Unit Size
11864614 10 Tubes Pack of 10
1 options
This item is not returnable. View return policy
Product Code. 11864614 Supplier Thermo Scientific™ Supplier No. 28601
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Reliably label (iodinate) proteins and peptides with iodine-125 using glass test tubes that are pre-coated (activated) with iodination reagent.
Thermo Scientific™ Pierce Iodination Tubes are 12 x 75mm glass test tubes that are pre-coated at the bottom with Pierce Iodination Reagent, which activates of iodine-125 for protein or peptide iodination procedures.
The popularity of Thermo Scientific Pierce Iodination Reagent stems from its gentle and highly efficient chemistry. We offer this reagent in the most convenient and versatile form on the market - pre-coating the inside surface of 12 mm x 75 mm test tubes.
Highlights:
  • Iodinations can be completed in less than 2 minutes
  • Eliminates the tedious reagent surface coating step
  • Provides a consistent reagent-coated and flake-resistant surface in the tube
  • Allows iodinations to be carried out directly in the tube
  • Enables 1252 1 to be pre-activated directly in the tube
  • Offers the opportunity to iodinate protein without ever having the protein contact the iodination reagent directly
TRUSTED_SUSTAINABILITY
Specifications
Product Type Iodination Tube
Quantity 10 Tubes
Content And Storage Upon receipt store at room temperature protected from moisture. After opening barrier pouch, close the interior resealable bag containing the tubes with the yellow closures and desiccant.
Product Line Pierce
Label or Dye Iodine-125
Label Type Radioisotope
For Research Use Only. Not for use in diagnostic procedures.