Learn More
Invitrogen™ EGFR Multispecies In-Cell ELISA Kit, Colorimetric
In-cell ELISA Kit
Marca:  Invitrogen™ 62205
Detalles adicionales : Peso : 0.01000kg
Código de producto. 11811194
  • 536.00€ / cada uno
Fecha de envío estimada
del proveedor 03-02-2020
Añadir a la cesta
Descripción
Descripción
The protein encoded by this gene is a transmembrane glycoprotein that is a member of the protein kinase superfamily. This protein is a receptor for members of the epidermal growth factor family. EGFR is a cell surface protein that binds to epidermal growth factor. Binding of the protein to a ligand induces receptor dimerization and tyrosine autophosphorylation and leads to cell proliferation. Mutations in this gene are associated with lung cancer. Multiple alternatively spliced transcript variants that encode different protein isoforms have been found for this gene.
Especificaciones
Especificaciones
P00533
Supernatant
HRP
Multi-component kit with HRP-conjugate
Blocking Buffer, Components included only in the target-specific kits:Antibody #1 and Antibody #2, Elution Buffer, 2 x 30mL, 20X Tris Buffered Saline, Surfact-Amps 20, Surfact-Amps X-100, HRP Conjugate, TMB Substrate, TMB Stop Solution, Janus Green Whole-Cell Stain, Elution Buffer, Thin Plate Seal Assembly
RUO
2°C to 8°C
1 hr. 20 min.
Human
HRP
96 Tests
Human
Pierce EGFR Colorimetric In-Cell ELISA Kit
Colorimetric Microplate Reader
ELISA Kit
Supernatant, 10,000 cells
96 microplate wells of overnight-cultured cells
24 hr.
EGFR