Learn More
Thermo Scientific™ BMH (bismaleimidohexane)
Product Code. 10581284 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity Unit Size
10581284 50 mg 50mg
1 options
This item is not returnable. View return policy
Product Code. 10581284 Supplier Thermo Scientific™ Supplier No. 22330
 Restricted Item
Fecha estimada de envoi: 03-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Crosslink sulfhydryls to sulfhydryls (cysteine residues in proteins or peptides) with this long crosslinker composed of maleimide groups and 11-atom (13.0A) spacer arm.
Thermo Scientific™ Pierce BMH is a long-arm, maleimide crosslinker for covalent, irreversible conjugation between sulfhydryl groups (e.g., protein or peptide cysteines).
Highlights:
  • Reactive groups: maleimide (homobifunctional)
  • Reactive toward: sulfhydryl groups
TRUSTED_SUSTAINABILITY
Specifications
Chemical Reactivity Sulfhydryl-Sulfhydryl
Cleavable No
Description BMH
Molecular Weight (g/mol) 276.29
PEGylated No
Spacer Arm Length 13.0 Å
Content And Storage Upon receipt store at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety Maleimide
Spacer Medium (10 to 30 Å)
Form Powder
Quantity 50 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less
compliance-icons
Product Identifier
  • BMH (bismaleimidohexane)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 1,1-(Hexane-1,6-diyl)bis-1H-pyrrole-2,5-dione
For Research Use Only. Not for use in diagnostic procedures.