Learn More
Thermo Scientific™ Ensayo de proteínas Pierce™ BCA™
Green-to-blue (A562nm), precise, detergent-compatible assay reagent to measure total protein concentration vs. protein standard.
66.00€ - 491.00€
Productos 7
Código de producto Marca Descripción Cantidad. Precio Cantidad y disponibilidad  
Código de producto Marca Descripción Cantidad. Precio Cantidad y disponibilidad  
10475944 Thermo Scientific™
23224
Reactivo B de ensayo de proteínas BCA Pierce 25 ml 66.00€
25 ml
En stock 104
Añadir a la cesta
Ver productos alternativos »
10678484 Thermo Scientific™
23227
Kit de ensayo de proteínas BCA Pierce Kit de 500 ml 144.00€
500 ml
En stock 308
Añadir a la cesta
10752735 SDS Thermo Scientific™
23222
Pierce BCA Protein Assay Reagent A 3.75L 491.00€
3.75 litro
Fecha de envío estimada
del proveedor 23-01-2019
Añadir a la cesta
10741395 Thermo Scientific™
23225
Kit de ensayo de proteínas BCA Pierce Kit de 1 l 221.00€
1 litro
En stock 390
Añadir a la cesta
11881345 SDS Thermo Scientific™
23223
Reactivo A de ensayo de proteínas con ácido bicinconínico (BCA) Pierce 1 l 174.00€
1000 ml
En stock 51
Añadir a la cesta
Ver productos alternativos »
10198543 SDS Thermo Scientific™
23221
Reactivo A de ensayo de proteínas con ácido bicinconínico (BCA) Pierce 250 ml 79.50€
250 ml
En stock 33
Añadir a la cesta
Ver productos alternativos »
10056623 SDS Thermo Scientific™
23228
Reactivo A de ensayo de proteínas con ácido bicinconínico (BCA) Pierce 500 ml 97.25€
500 ml
En stock 160
Añadir a la cesta
Ver productos alternativos »
Descripción
Descripción
Used in more labs than any other detergent-compatible protein assay, Pierce BCA Reagents provide accurate determination of protein concentration with most sample types encountered in protein research. The Pierce BCA Assay can be used to assess yields in whole cell lysates and affinity-column fractions, as well as to monitor protein contamination in industrial applications. Compared to most dye-binding methods, the BCA Assay is affected much less by protein compositional differences, providing greater protein-to-protein uniformity.
Highlights:
  • Colorimetric – estimate visually or measure with a standard spectrophotometer or plate reader (562nm)
  • Excellent uniformity – exhibits less protein-to-protein variation than dye-binding methods
  • Compatible – unaffected by typical concentrations of most ionic and nonionic detergents
  • Moderately fast – much easier and four times faster than the classical Lowry method
  • High linearity – linear working range for BSA equals 20 to 2000µg/mL
  • Sensitive – detect down to 5µg/mL with the enhanced protocol
BCA Protein Assay Applications:
  • Studying protein:protein interactions
  • Measuring column fractions after affinity chromatography
  • Estimating percent recovery of membrane proteins from cell extracts
  • High-throughput screening of fusion proteins