Learn More
Uvex™ Phynomic Wet Plus Safety Glove
The Uvex phynomic series is ideal for all types of work where a high level of feeling is required.
Marca: Uvex™ 6006109
Detalles adicionales : Peso : 0.04560kg
Descripción
- Safety glove with water-repellent aqua-polymer foam coating for use outdoors
- Aqua-polymer foam coating on palm, fingertips and parts of the back of the hand
- Outstanding mechanical abrasion resistance thanks to the durable coating
- Good grip in damp, wet and oily areas
- High level of breathability with the aqua-polymer foam coating
- Good tactile feel when assembling parts
- Perfect fit with 3D Ergo technology
- Excellent skin tolerance, confirmed by dermatologists (uvex pure Standard)
- Free of harmful substances in line with OEKO-Tex Standard 100
- Excellent skin tolerance – confirmed by the proDERM Institute for Applied Dermatological Research (proDerm studies: 11.0356-02, 11.0482-11)
Especificaciones
EN 388:2016 (3 1 3 1 X) | |
Anthracite, Blue | |
9 |
Aqua-Polymer Foam | |
Elastane, Polyamide |