Learn More
Uvex™ Phynomic Wet Plus Safety Glove
The Uvex phynomic series is ideal for all types of work where a high level of feeling is required.
Marca:  Uvex™ 6006109
Detalles adicionales : Peso : 0.04560kg
 Ver más versiones de este producto
Código de producto. 17797579
  • 44.85€ / 10 pares
Fecha estimada de envío: 05-04-2023
para ver el stock
Añadir a la cesta
Descripción
Descripción
  • Safety glove with water-repellent aqua-polymer foam coating for use outdoors
  • Aqua-polymer foam coating on palm, fingertips and parts of the back of the hand
  • Outstanding mechanical abrasion resistance thanks to the durable coating
  • Good grip in damp, wet and oily areas
  • High level of breathability with the aqua-polymer foam coating
  • Good tactile feel when assembling parts
  • Perfect fit with 3D Ergo technology
  • Excellent skin tolerance, confirmed by dermatologists (uvex pure Standard)
  • Free of harmful substances in line with OEKO-Tex Standard 100
  • Excellent skin tolerance – confirmed by the proDERM Institute for Applied Dermatological Research (proDerm studies: 11.0356-02, 11.0482-11)
Especificaciones
Especificaciones
EN 388:2016 (3 1 3 1 X)
Anthracite, Blue
9
Aqua-Polymer Foam
Elastane, Polyamide