Learn More
Uvex™ Phynomic Pro 2 Safety Glove
The Uvex phynomic pro 2 is a sensitive safety glove for assembly activities in slightly damp/oily areas.
Marca: Uvex™ 6006412
Detalles adicionales : Peso : 0.05800kg
Descripción
- Innovative bamboo-fiber assembly gloves
- Dirt- and moisture-repellent aqua-polymer pro coating on palm and fingertips
- Upper made of bamboo, polyamide and elastane
- Good grip in dry and slightly damp/oily areas
- Excellent impermeability and breathability when working with resins and highly viscous pastes
- Safety glove free from allergenic accelerators – health protection and skin compatibility dermatologically tested (proDERM)
- Suitable for allergy sufferers and as a solvent-free alternative to PU gloves
- Harmless according to OEKO-TEX Standard 100
- Light and breathable safety glove
- Safety glove with ergonomic fit
- Excellent wearer comfort thanks to skin-friendly bamboo fibers in the lining
Especificaciones
EN 388 (2 1 2 1 X) | |
Anthracite, Blue | |
12 |
Aqua-Polymer | |
Bamboo, Elastane, Polyamide |