Learn More
Uvex™ Phynomic Pro 2 Safety Glove
The Uvex phynomic pro 2 is a sensitive safety glove for assembly activities in slightly damp/oily areas.
Marca:  Uvex™ 6006412
Detalles adicionales : Peso : 0.05800kg
 Ver más versiones de este producto
Código de producto. 17797589
  • 47.75€ / 10 pares
Fecha estimada de envío: 05-04-2023
para ver el stock
Añadir a la cesta
Descripción
Descripción
  • Innovative bamboo-fiber assembly gloves
  • Dirt- and moisture-repellent aqua-polymer pro coating on palm and fingertips
  • Upper made of bamboo, polyamide and elastane
  • Good grip in dry and slightly damp/oily areas
  • Excellent impermeability and breathability when working with resins and highly viscous pastes
  • Safety glove free from allergenic accelerators – health protection and skin compatibility dermatologically tested (proDERM)
  • Suitable for allergy sufferers and as a solvent-free alternative to PU gloves
  • Harmless according to OEKO-TEX Standard 100
  • Light and breathable safety glove
  • Safety glove with ergonomic fit
  • Excellent wearer comfort thanks to skin-friendly bamboo fibers in the lining
Especificaciones
Especificaciones
EN 388 (2 1 2 1 X)
Anthracite, Blue
12
Aqua-Polymer
Bamboo, Elastane, Polyamide