Learn More
Uvex™ Phynomic AirLite C ESD Safety Glove
The Uvex phynomic airLite ESD is a light safety glove that offers a noticeable difference in wearer comfort.
Marca: Uvex™ 6008407
Detalles adicionales : Peso : 0.04370kg
Descripción
- The lightest and most sensitive uvex phynomic cut protection glove in Cut Level C
- Outer material made from innovative Dyneema Diamond 2.0 cut protection fibers, polyamide, elastane and carbon
- Breathable 'airLite' aqua-polymer coating on palm and fingertips
- Safety glove free from glass and steel fibers
- Safety glove with touchscreen function – for use on almost all screens, tablets and mobile phones
- Safety glove with ESD function in line with DIN EN 16350:2014 – protects workpiece against electrostatic charge
- High cut protection (Level C)
- Good grip in dry and slightly damp areas
- Free from accelerators, and health protection and skin compatibility dermatologically approved (proDERM)
- Very lightweight and breathable safety glove
- Outstanding sensitivity thanks to new breathable 'airLite' aqua-polymer coating combined with fine liner (18 gauge)
- Safety gloves with knitted cuff and ergonomic fit
Especificaciones
EN 388 (3 X 4 2 C) and DIN EN 16350:2014 | |
Black, Blue | |
7 |
Aqua-Polymer Foam | |
Carbon, Elastane, Polyamide |