Learn More
Phosphorus, red, 97%, pure, powder
Product Code. 10369043 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 kg
Unit Size:
2.5kg
Product Code. Quantity Unit Size
10369043 2.5 kg 2.5kg
1 options
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 10369043 Supplier Thermo Scientific Acros Supplier No. 345970025
 Restricted Item
En stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
CAS: 7723-14-0 | P | 30.97 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7723-14-0
Molecular Formula P
Molecular Weight (g/mol) 30.97
MDL Number MFCD00133771
InChI Key OAICVXFJPJFONN-UHFFFAOYSA-N
IUPAC Name phosphorus
SMILES [P]
Specifications
Melting Point 590.0°C
Quantity 2.5 kg
Assay Percent Range 97%
Linear Formula P
Fieser 01,861
Merck Index 15, 7459
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in absolute alcohol, insoluble in cs2, ether, nh3
Formula Weight 30.96
Grade Pure
Packaging Glass bottle
Chemical Name or Material Phosphorus, red
Show More Show Less
compliance-icons
Product Identifier
  • Phosphorus, red
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable solid Category 1
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.