missing translation for 'onlineSavingsMsg'
Learn More

PHOSPHO1 Antibody (011), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-90192AF647

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214769

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

PHOSPHO1 Monoclonal specifically detects PHOSPHO1 in Human samples. It is validated for ELISA.
Especificaciones

Especificaciones

PHOSPHO1
Monoclonal
Alexa Fluor 647
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human
Purified
ELISA
11
ELISA
EC 3.1.3.75, phosphatase, orphan 1, phosphoethanolamine/phosphocholine phosphatase
This antibody was obtained from a rabbit immunized with purified, recombinant Human PHOSPHO1 (Q8TCT1-1; Met1-Cys267).
0.1 mL
Protein Phosphatase
162466
Store at 4°C in the dark.
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales