missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Stat4 (Tyr693) Rabbit anti-Human, Mouse, PE, Clone: Stat4Y693-F6, Invitrogen™
Rabbit Monoclonal Antibody
Marca: Thermo Scientific MA537332
Detalles adicionales : Peso : 0.01000kg
Descripción
Phospho-Stat4 (Tyr693) Monoclonal antibody specifically detects Phospho-Stat4 (Tyr693) in Human, Mouse samples. It is validated for Flow CytometryEspecificaciones
Phospho-Stat4 (Tyr693) | |
Monoclonal | |
PE | |
STAT4 | |
CIS4; HSPC060; OTTHUMP00000163553; Signal transducer and activator of transcription 4; SLEB11; SOCS4; SOCS6; SSI4; STAI4; STAT; Stat4; STATI4 | |
A synthetic phospho-peptide corresponding to residues surrounding Tyr693 of human phospho Stat4. | |
10 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
Stat4Y693-F6 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
P42228, Q14765 | |
Rabbit | |
Protein A/G | |
RUO | |
20849, 6775 | |
2-8°C | |
Liquid |