missing translation for 'onlineSavingsMsg'
Learn More

Phospho-RARA/RARB (Ser96) Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA564622

Detalles adicionales : Peso : 0.01000kg

Código de producto. 15762686

  • 472.00€ / 100 microlitros
Fecha estimada de envío: 22-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Phospho-RARA/RARB (Ser96) Polyclonal Antibody detects endogenous levels of RARA/RARB only when phosphorylated at Ser96.

Phospho-RARA/RARB (Ser96) Polyclonal antibody specifically detects Phospho-RARA/RARB (Ser96) in Human, Mouse, Rat samples. It is validated for Western Blot.
Especificaciones

Especificaciones

Phospho-RARA/RARB (Ser96)
Polyclonal
Unconjugated
RARB
A830025K23; HAP; HBV-activated protein; hepatitis B virus activated protein; MCOPS12; Nr1b1; Nr1b2; Nuclear receptor subfamily 1 group B member 1; nuclear receptor subfamily 1 group B member 2; nucleophosmin-retinoic acid receptor alpha fusion protein NPM-RAR long form; RAR; RAR alpha 1; RARA; RAR-alpha; RARalpha1; Rarb; RAR-beta; RARbeta1; RAR-epsilon; retinoic acid; retinoic acid nuclear receptor alpha variant 1; retinoic acid nuclear receptor alpha variant 2; retinoic acid receptor; retinoic acid receptor alpha; Retinoic acid receptor beta; retinoic acid receptor, alpha; retinoic acid receptor, alpha polypeptide; retinoic acid receptor, beta; retinoic acid receptor, beta polypeptide; RRB2
Rabbit
Affinity chromatography
RUO
19401, 218772, 24705, 24706, 5914, 5915
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P10276, P10826, P11416, P22605
RARA, RARB
A synthesized peptide derived from human Retinoic Acid Receptor alpha/beta (Phospho-Ser96)
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales