missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PLK1 (Thr210) Recombinant Rabbit Monoclonal Antibody (PLK1T210-C2), APC, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537127
Detalles adicionales : Peso : 0.01000kg
Descripción
Phospho-PLK1 (Thr210) Recombinant Monoclonal antibody specifically detects Phospho-PLK1 (Thr210) in Human samples. It is validated for Flow Cytometry.Especificaciones
Phospho-PLK1 (Thr210) | |
Monoclonal | |
APC | |
Plk1 | |
cell cycle regulated protein kinase; Plk; PLK 1; Plk1; PLK-1; plk1.S; plx1; polo (Drosophia)-like kinase; polo like kinase; polo like kinase 1; polo like kinase 1 S homeolog; polo-like kinase; polo-like kinase 1; polo-like kinase homolog; polo-like protein kinase; serine/threonine-protein kinase 13; Serine/threonine-protein kinase PLK1; STPK13; STPK14; Unknown (protein for MGC:133927); XELAEV_18047692mg | |
Rabbit | |
Protein A/G | |
RUO | |
5347 | |
4° C | |
Liquid |
Flow Cytometry | |
PLK1T210-C2 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
P53350 | |
Plk1 | |
A synthetic phospho-peptide corresponding to residues surrounding Thr210 of human phospho PLK1. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG κ |