missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MAPK9 (Thr183, Tyr185) Recombinant Rabbit Monoclonal Antibody (SAPKT183Y185-A11), PE, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537379
Detalles adicionales : Peso : 0.01000kg
Descripción
Phospho-MAPK9 (Thr183, Tyr185) Recombinant Monoclonal antibody specifically detects Phospho-MAPK9 (Thr183, Tyr185) in Human samples. It is validated for Flow Cytometry.Especificaciones
Phospho-MAPK9 (Thr183, Tyr185) | |
Monoclonal | |
PE | |
MAPK9 | |
AI851083; c-Jun kinase 2; c-Jun N-terminal kinase 2; I79_012561; JNK; JNK/SAPK alpha; JNK1; JNK1A2; JNK2; JNK21B1/2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; JNK-46; JNK-55; Jun kinase; MAP kinase 10; MAP kinase 8; MAP kinase 9; MAPK 10; MAPK 9; MAPK10; MAPK8; Mapk9; mitogen activated protein kinase 9; mitogen-activated protein kinase 9; OTTHUMP00000161542; OTTHUMP00000161543; p54a; p54-alpha; p54aSAPK; PRKM8; PRKM9; protein kinase, mitogen-activated 9; SAPK; SAPK1; SAPK1a; SAPK1c; SAPK-alpha; stress activated protein kinase alpha II; stress-activated protein kinase 1a; Stress-activated protein kinase JNK2 | |
Rabbit | |
Protein A/G | |
RUO | |
5601 | |
4° C | |
Liquid |
Flow Cytometry | |
SAPKT183Y185-A11 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
P45984 | |
MAPK9 | |
A synthetic phospho-peptide corresponding to residues surrounding Thr183/Tyr185 of human phospho SAPK/JNK. | |
10 Tests | |
Primary | |
Human | |
Antibody | |
IgG κ |