missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Lck (Tyr505) Recombinant Rabbit Monoclonal Antibody (LckY505-A3), PE, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537341
Detalles adicionales : Peso : 0.01000kg
Descripción
Phospho-Lck (Tyr505) Recombinant Monoclonal antibody specifically detects Phospho-Lck (Tyr505) in Human samples. It is validated for Flow Cytometry.Especificaciones
Phospho-Lck (Tyr505) | |
Monoclonal | |
PE | |
LCK | |
EC 2.7.10.2; Hck-3; IMD22; kinase Lck; Lck; LCK proto-oncogene, Src family tyrosine kinase; Lck1; Lcktkr; leukocyte C-terminal Src kinase; LSK; Lskt; Lsk-t; Lymphocyte cell-specific protein-tyrosine kinase; lymphocyte protein tyrosine kinase; lymphocyte-specific protein tyrosine kinase; OTTHUMP00000008640; OTTHUMP00000008740; OTTHUMP00000008741; p56(LSTRA) protein-tyrosine kinase; p56<lck>; p56lck; p56-LCK; pp58lck; Protein YT16; Proto-oncogene Lck; Proto-oncogene tyrosine-protein kinase LCK; RP4-675E8.4; t cell-specific protein-tyrosine kinase; T-lymphocyte specific protein tyrosine kinase p56lck; tyr; tyrosine-protein kinase Lck; YT16 | |
Rabbit | |
Protein A/G | |
RUO | |
3932 | |
4° C | |
Liquid |
Flow Cytometry | |
LckY505-A3 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
P06239 | |
LCK | |
A synthetic phospho-peptide corresponding to residues surrounding Tyr505 of human phospho Lck. | |
10 Tests | |
Primary | |
Human | |
Antibody | |
IgG κ |