missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-KIF20A (Ser528) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA540224
Detalles adicionales : Peso : 0.00910kg
Descripción
Phospho-KIF20A (Ser528) Polyclonal antibody specifically detects Phospho-KIF20A (Ser528) in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Especificaciones
Phospho-KIF20A (Ser528) | |
Polyclonal | |
Unconjugated | |
KIF20A | |
AA415432; GG10_2; KIF20A; kinesin family member 20A; kinesin-like protein 174; Kinesin-like protein KIF20A; mitotic kinesin-like protein 2; MKlp2; RAB6 interacting, kinesin-like (rabkinesin6); rab6-interacting kinesin-like protein; RAB6KIFL; Rabkinesin-6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10112 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O95235 | |
KIF20A | |
Synthesized peptide derived from human KIF20A around the phosphorylation site of S528. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |