missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-DAPK1 (Ser289) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA564573
Detalles adicionales : Peso : 0.01000kg
Descripción
Phospho-DAPK1 (Ser289) Polyclonal Antibody detects endogenous levels of DAPK1 only when phosphorylated at Ser289.
Phospho-DAPK1 (Ser289) Polyclonal antibody specifically detects Phospho-DAPK1 (Ser289) in Human, Mouse samples. It is validated for Western Blot.Especificaciones
Phospho-DAPK1 (Ser289) | |
Polyclonal | |
Unconjugated | |
DAPK1 | |
D13Ucla1; DAP kinase 1; DAPK; DAPK1; DAP-Kinase; death associated protein kinase 1; death-associated protein kinase 1; DKFZp781I035; OTTHUMP00000216193 | |
Rabbit | |
Affinity chromatography | |
RUO | |
1612, 69635 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P53355, Q80YE7 | |
DAPK1 | |
A synthesized peptide derived from human DAPK1 (Phospho-Ser289) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |