missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-c-Met (Tyr1313) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA564802
Detalles adicionales : Peso : 0.01000kg
Descripción
Phospho-c-Met (Tyr1313) Polyclonal Antibody detects endogenous levels of c-Met only when phosphorylated at Tyr1313.
Phospho-c-Met (Tyr1313) Polyclonal antibody specifically detects Phospho-c-Met (Tyr1313) in Human, Mouse, Rat samples. It is validated for Western Blot.Especificaciones
Phospho-c-Met (Tyr1313) | |
Polyclonal | |
Unconjugated | |
MET | |
AI838057; AUTS9; bHLHe59; CG1705; CG1705-PA; CG1705-PB; cmet; c-met; c-met proto-oncogene; c-Met receptor tyrosine kinase; c-Met/HGF receptor; D249; DFNB97; Dmel\CG1705; Dmel_CG1705; DmMet; EC 2.7.10.1; Hepatocyte growth factor receptor; HGF; HGF receptor; HGF receptor c-Met; HGF/SF receptor; HGFR; HGF-SF receptor; juvenile hormone resistance; met; met proto-oncogene; met proto-oncogene (hepatocyte growth factor receptor); met proto-oncogene tyrosine kinase; MET proto-oncogene, receptor tyrosine kinase; Met/Met1; Met; protein tyrosin kinase; methoprene tolerant; Methoprene-tolerant; Methoprene-tolerant protein; Met-PA; Met-PB; Mett; oncogene MET; Par4; Proto-oncogene c-Met; RCCP2; resistance (1) juvenile H; resistance to juvenile hormone; Rst(1)JH; sc MET; scatter factor receptor; SF receptor; soluble c met; tyrosine-protein kinase Met | |
Rabbit | |
Affinity chromatography | |
RUO | |
17295, 24553, 4233 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P08581, P16056, P97523 | |
MET | |
A synthesized peptide derived from human Met (Phospho-Tyr1313) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |