missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-c-Fos (Ser32) Recombinant Rabbit Monoclonal Antibody (cFosS32-BA9), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537173
Detalles adicionales : Peso : 0.01000kg
Descripción
Phospho-c-Fos (Ser32) Recombinant Monoclonal antibody specifically detects Phospho-c-Fos (Ser32) in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Especificaciones
Phospho-c-Fos (Ser32) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.10% BSA, 50% glycerol and 0.02% sodium azide | |
P01100, P01101, P12841 | |
FOS | |
A synthetic phospho-peptide corresponding to residues surrounding Ser32 of human phospho c-Fos. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG κ |
Flow Cytometry | |
cFosS32-BA9 | |
Unconjugated | |
FOS | |
activator protein 1; AP-1; cellular oncogene c-fos; cellular oncogene fos; cFos; C-FOS; c-fos oncogene; c-fos protooncogene protein; D12Rfj1; FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS); FBJ murine osteosarcoma viral oncogene homolog; FBJ murine osteosarcoma viral oncogene homolog B; FBJ osteosarcoma oncogene; FBJ osteosarcoma oncogene B; FBJ osteosarcoma viral oncogene isoform deltaFosb-2; FOS; Fos proto-oncogene, AP-1 trancription factor subunit; Fos proto-oncogene, AP-1 transcription factor subunit; Fosb; FosB proto-oncogene, AP-1 transcription factor subunit; fra-2; G0/G1 switch regulatory protein 7; G0S7; I79_009428; p55; protein fosB; proto-oncogene cFOS; proto-oncogene c-Fos; proto-oncogene protein c-fos; v-fos FBJ murine osteosarcoma viral oncogene homolog | |
Rabbit | |
Protein A/G | |
RUO | |
14281, 2353, 314322 | |
-20°C | |
Liquid |