missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Btk (Tyr551) Recombinant Rabbit Monoclonal Antibody (BtkY551-G12), Biotin, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537460
Detalles adicionales : Peso : 0.01000kg
Descripción
Phospho-Btk (Tyr551) Recombinant Monoclonal antibody specifically detects Phospho-Btk (Tyr551) in Human samples. It is validated for Flow Cytometry.Especificaciones
Phospho-Btk (Tyr551) | |
Monoclonal | |
Biotin | |
BTK | |
agammaglobulinaemia tyrosine kinase; agammaglobulinemia tyrosine kinase; AGMX1; AI528679; AT; ATK; B-cell progenitor kinase; BPK; Bruton agammaglobulinemia tyrosine kinase; Bruton tyrosine kinase; Bruton's tyrosine kinase; BTK; dominant-negative kinase-deficient Brutons tyrosine kinase; IMD1; Kinase EMB; MGC126261; MGC126262; OTTHUMP00000023677; PSCTK1; RP1-164F3.2; truncated Bruton agammaglobulinemia tyrosine kinase; tyrosine-protein kinase BTK; Tyrosine-protein kinase BTK (Brutons tyrosine kinase) (Agammaglobulinaemia tyrosine kinase) (ATK) (B cell progenitor kinase) (BPK) (Kinase EMB); tyrosine-protein kinase BTK isoform (lacking exon 13 to 17); tyrosine-protein kinase BTK isoform (lacking exon 14); xid; XLA; X-linked immune deficiency | |
Rabbit | |
Protein A/G | |
RUO | |
695 | |
4° C | |
Liquid |
Flow Cytometry | |
BtkY551-G12 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
Q06187 | |
BTK | |
A synthetic phospho-peptide corresponding to residues surrounding human Btk (Y551). | |
10 Tests | |
Primary | |
Human | |
Antibody | |
IgG κ |