Learn More
PHF10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA563154
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: GRGDEKRKNK GTSDSSSGNV SEGESPPDSQ EDSFQGRQKS KDKAATPRKD GPKRSVLSKS VPGYKPKVI Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 93%.
This gene contains a predicted ORF that encodes a protein with two zinc finger domains. The function of the encoded protein is not known. Sequence analysis suggests that multiple alternatively spliced transcript variants are derived from this gene but the full-length nature of only two of them is known. These two splice variants encode different isoforms. A pseudogene for this gene is located on Xq28.Especificaciones
PHF10 | |
Polyclonal | |
Unconjugated | |
PHF10 | |
1810055P05Rik; AV024533; BAF45A; BRG1-associated factor 45a; BRG1-associated factor, 45-KD, A; PHD finger protein 10; PHD finger protein 10, isoform a; PHD zinc finger protein XAP135; Phf10; XAP135 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55274 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WUB8 | |
PHF10 | |
Recombinant protein corresponding to Human PHF10 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only