Learn More
Acrilato de fenilo, 97 %, Thermo Scientific Chemicals
Catalog Number p-7058378
Click to view available options
Cantidad:
5 g
25 g
937-41-7
C9H8O2
148.161
MFCD00048145
WRAQQYDMVSCOTE-UHFFFAOYSA-N
phenyl acrylate, phenylacrylate, 2-propenoic acid, phenyl ester, phenol acrylate, phenyl-acrylate, acrylic acid phenyl ester, phenyl acrylate 5g, 2-propenoic acid,phenyl ester, acrylic acid phenyl ester, stabilized with bht
61242
prop-2-enoato de fenilo
C=CC(=O)OC1=CC=CC=C1
937-41-7
C9H8O2
148.161
MFCD00048145
WRAQQYDMVSCOTE-UHFFFAOYSA-N
phenyl acrylate, phenylacrylate, 2-propenoic acid, phenyl ester, phenol acrylate, phenyl-acrylate, acrylic acid phenyl ester, phenyl acrylate 5g, 2-propenoic acid,phenyl ester, acrylic acid phenyl ester, stabilized with bht
61242
prop-2-enoato de fenilo
C=CC(=O)OC1=CC=CC=C1
Phenyl acrylate reacts with benzaldehyde to produce 2-(hydroxy-phenyl-methyl)-acrylic acid phenyl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
El fenilacrilato reacciona con el benzaldehído para producir éster fenílico del ácido 2-(hidroxi-fenil-metil)-acrílico.
Solubilidad
Insoluble en agua.
Notas
Incompatible con agentes oxidantes fuertes. Almacenar a 4 °C. Mantener el recipiente bien cerrado. Almacenar en un lugar fresco y seco y en recipientes bien sellados.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Phenyl acrylate
Densidad 1.076
Punto de ebullición 89°C to 90°C (12 mmHg)
Índice de refracción 1.521
Cantidad 5 g
Número UN UN3082
Beilstein 2041124
Información de solubilidad Insoluble in water.
Formula Weight (peso de la fórmula) 148.16
Porcentaje de pureza 97%