missing translation for 'onlineSavingsMsg'
Learn More

Fenol, ACS, +99 %, estabilizado, Thermo Scientific Chemicals

Catalog Number p-7050000
29.33 EUR hasta el 2024-03-29
Use el código promocional "21615" para aplicar la oferta.


Click to view available options
Cantidad:
100 g
500 g
2 kg
Este artículo no se puede devolver. Vea la política de devoluciones
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:21615
108-95-2
C6H6O
94.11
MFCD00002143
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
996
CHEBI:15882
fenol
OC1=CC=CC=C1
Este artículo no se puede devolver. Vea la política de devoluciones
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:21615
108-95-2
C6H6O
94.11
MFCD00002143
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
996
CHEBI:15882
fenol
OC1=CC=CC=C1

Phenol is used as a precursor for cyclohexanone, plastics, nonionic detergents and pharmaceutical drugs like aspirin. It acts as an anesthetic in chloraseptic. It reacts with acetone to get bisphenol-A which is used in the synthesis of poly carbonates and epoxide resins. It is also used in the manufacture of synthetic resins, dyes, pharmaceuticals, synthetic tanning agents, perfumes, lubricating oils and solvents. In molecular biology, it is used in the extraction of nucleic acid from tissues by using the liquid/liquid phenol-chloroform extraction technique. It is an active component of paint strippers which are used for the removal of epoxy and polyurethane. It is also used in the preparation of cosmetics, hair colors and skin lightening preparations. In the field of medicine, its spray is useful in helping sore throat.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Phenol
Nombre de nota Stabilized
Punto de fusión 40°C to 43°C
Densidad 1.071 g/mL
Punto de ebullición 181°C
Temperatura de inflamación 79°C (175°F)
Olor Sweet
Cantidad 100 g
Número UN UN1671
Beilstein 969616
Sensibilidad Air Sensitive
Índice Merck 14,7241
Información de solubilidad Soluble in water,alcohol,chloroform,ether,benzene,glycerol,acetone,carbon disulfide and aqueous alkali hydroxides.
Formula Weight (peso de la fórmula) 94.11
Porcentaje de pureza ≥99%
Grado ACS
Forma física cristalino/a/s
Mostrar más Mostrar menos

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'