Learn More
Esculape Laboratoire™ Pharmacy Cabinets
Product Code. 11738423
Change view
Click to view available options
Product Type:
Pharmacy Cabinet - Empty
Pharmacy Cabinet - Filled
Height (Metric):
14 cm
20 cm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Height (Metric) Unit Size
11738423 Pharmacy Cabinet - Filled 20 cm Each
11718423 Pharmacy Cabinet - Empty 14 cm Each
11708423 Pharmacy Cabinet - Filled 14 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11738423 Supplier Esculape Laboratoire™ Supplier No. 711 03 04
  • ONLINE EXCLUSIVE PRICE
    495.60€
    276.11€ / paquete de 1
    Ahorro 219.49€
    44% Descuento
Fecha estimada de envoi: 01-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Depth (Metric) 53 cm
For Use With (Application) To accommodate all the first aid equipment, offices, workshops and laboratories
Length (Metric) 53 cm
Product Type Pharmacy Cabinet - Filled
Quantity 1 Ea.
Height (Metric) 20 cm