Learn More
Macherey-Nagel™ pH-Fix pH Indicator Strips
Product Code. 15364184 Shop All Macherey-Nagel Products
Change view
Click to view available options
pH Range:
0 to 14
1.7 to 3.8
2 to 9
Unit Size:
100 units
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
15364184 2 to 9 100 units
15705802 0 to 14 Pack of 100
15324194 1.7 to 3.8 100 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15364184 Supplier Macherey-Nagel™ Supplier No. 92118
  • ONLINE EXCLUSIVE PRICE
    20.30€
    16.10€ / paquete de 100
    Ahorro 4.20€
    20% Descuento
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Highly appreciated standard strips for many pH testing applications.
  • Easy dip and read procedure provides reliable results in seconds
  • Due to the patented pH-Fix technology, the indicator is chemically bound to the test pad
  • Dye does not wash out, so clothes stay clean and samples remain pure
Specifications
pH Range 2 to 9
Format Strip
Quantity 100 Strips
Type pH Indicator Paper
Length (Metric) 85 mm
Width (Metric) 6 mm