Learn More
Savillex™ PFA Digestion Vessels
Product Code. 15296158
Change view
Click to view available options
Capacity (Metric):
60 mL
120 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15296158 120 mL Each
11912195 60 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15296158 Supplier Savillex™ Supplier No. 30012003
  • ONLINE EXCLUSIVE PRICE
    265.03€
    131.17€ / paquete de 1
    Ahorro 133.86€
    50% Descuento
 Restricted Item
Fecha estimada de envoi: 18-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Product Type Digestion Vessel
Capacity (Metric) 120 mL
For Use With (Application) Rapid sample digestion via microwave techniques
Material PFA