missing translation for 'onlineSavingsMsg'
Learn More

Peripheral Node Addressin Antibody (MECA-79R), DyLight 594, Novus Biologicals™

Rat Monoclonal Antibody

Marca:  Bio-Techne NBP2-78792DL594

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215647

  • 344.48€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

Peripheral Node Addressin Monoclonal specifically detects Peripheral Node Addressin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Especificaciones

Especificaciones

Peripheral Node Addressin
Monoclonal
DyLight 594
50mM Sodium Borate with 0.05% Sodium Azide
Rat
Protein A or G purified
RUO
Human, Mouse
IgG1
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
MECA-79R
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
MECA-79, Peripheral Node Addressin, PNAd
Mouse lymph node stromal cells
0.1 mL
Primary
Store at 4°C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales