missing translation for 'onlineSavingsMsg'
Learn More

Perilipin-2/ADFP Antibody (ADFP/1494), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-54598AF350

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215723

  • 569.10€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Perilipin-2/ADFP Monoclonal specifically detects Perilipin-2/ADFP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

Perilipin-2/ADFP
Monoclonal
Alexa Fluor 350
50mM Sodium Borate with 0.05% Sodium Azide
PLIN2
Recombinant fragment (around aa 249-376) of human Perilipin-2/ADFP protein (exact sequence is proprietary) (Uniprot: Q99541)
0.1 mL
Cancer, Hypoxia, Lipid and Metabolism, Lipid Droplets, Signal Transduction
Recognizes a protein of 48kDa, which is identified as Adipophilin. It belongs to the perilipin family, members of which coat intracellular lipid storage droplets. This protein is associated with the lipid globule surface membrane material, and maybe involved in development and maintenance of adipose tissue. However, it is not restricted to adipocytes as previously thought, but is found in a wide range of cultured cell lines, including fibroblasts, endothelial and epithelial cells, and tissues, such as lactating mammary gland, adrenal cortex, Sertoli and Leydig cells, and hepatocytes in alcoholic liver cirrhosis, suggesting that it may serve as a marker of lipid accumulation in diverse cell types and diseases.
Store at 4°C in the dark.
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
ADFP/1494
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence
ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2
Mouse
Protein A or G purified
RUO
Primary
Human
IgG2b κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales