missing translation for 'onlineSavingsMsg'
Learn More

Perforin Antibody (PRF1/2470), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-79886AF700

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213546

  • 578.82€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Perforin Monoclonal specifically detects Perforin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

Perforin
Monoclonal
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
PRF1
Recombinant human Perforin protein fragment (around aa 413-552) (exact sequence is proprietary) (Uniprot: P14222)
0.1 mL
Adaptive Immunity, Apoptosis, Immunology, Innate Immunity
Human
IgG2c κ
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
PRF1/2470
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFPcytolysin
Mouse
Protein A or G purified
RUO
Primary
Store at 4°C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales