Learn More
Pentamethylenebis(magnesium bromide), 0.5M solution in THF, AcroSeal™
Product Code. 17797727 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
Each
Product Code. Quantity Packaging Unit Size
17797727 100 mL AcroSeal™ Glass Bottle Each
1 options
This item is not returnable. View return policy
Product Code. 17797727 Supplier Thermo Scientific Supplier No. 436001000
  • ONLINE EXCLUSIVE PRICE
    1492.86€
    1012.03€ / paquete de 1
    Ahorro 480.83€
    32% Descuento
 Restricted Item
Fecha estimada de envoi: 19-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
CAS: 23708-48-7 | C5H10Br2Mg2 | 278.55 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 23708-48-7, 109-99-9
Color Brown to Yellow or Gray
Density 0.9990g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C5H10Br2Mg2
MDL Number MFCD00012384
Quantity 100 mL
Specific Gravity 0.999
Synonym pentamethylenebis magnesium bromide, bromo 5-bromomagnesio pentyl magnesium, pentane-1,5-di magnesium bromide, pentamethylenebis magnesium bromide solution, pentamethylenebis magnesium bromide 0,5m w/w in thf, pentamethylenebis magnesium bromide solution, 0.5 m in thf, grignard reagent
Solubility Information Solubility in water: reacts.
InChI Key OUEGHZYBWJTUAD-UHFFFAOYSA-L
SMILES Br[Mg]CCCCC[Mg]Br
IUPAC Name dimagnesium;pentane;dibromide
Molecular Weight (g/mol) 278.55
PubChem CID 3637391
Formula Weight 278.55
Physical Form Solution
Chemical Name or Material Pentamethylenebis(magnesium bromide)